missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bio-11-UTP (75 mM)

Ambion Bio-11-UTP 75 mM (Biotinylated UTP) is intended for use as substrates in in vitro transcription reactions.

Brand:  Invitrogen™ AM8451

Code : 50

Additional Details : Weight : 0.00300kg

Product Code. 10269614

  • £1710.00 / Each
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Ambion Bio-11-UTP 75 mM (Biotinylated UTP) is intended for use as substrates in in vitro transcription reactions. These modified nucleotides can be utilized by a variety of RNA polymerases, including T7, T3, and SP6 RNA polymerases, as well as other RNA modifying enzymes. It is provided in one tube containing 30 µL (2,250 nmol).

Applications
Biotinylated RNA can be used in place of radioactively labeled RNA in many applications, including cRNA amplification and labeling for microarray target preparation. Detection is then carried out via one of a variety of streptavidin-based methods.

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Nucleotides
Dry Ice
Biotin
Ambion™
Liquid
Bio-11-UTP, 75 mM (30 μL)

Store at –70°C.
75 mM
30 μL
Nucleic Acid Labeling, Oligo Synthesis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.