Learn More
Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity
Description
Includes
AccuPrime™ Taq DNA Polymerase High Fidelity (40μL, at 5U/μL)) is supplied with: 50mM MgSO4 (1mL), 10X AccuPrime™ PCR Buffer I (1mL), 10X AccuPrime™ PCR Buffer II (1mL)
Buffer I and II differ in their concentration of thermostable AccuPrime™ protein. Their components are: 600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4, 20mM MgSO4, 2mM dGTP, 2mM dATP, 2mM dTTP, 2mM dCTP, thermostable AccuPrime™ protein, 10% glycerol. The supplied PCR Buffer is a 10X concentrate and should be diluted for use. Buffer I is optimized for plasmids, cDNA, and λDNA. Buffer II is optimized for genomic DNA.
Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates. It includes an enzyme blend of Platinum Taq DNA Polymerase and a proofreading enzyme, plus AccuPrime accessory proteins for improved PCR fidelity, yield, and specificity over other hot-start DNA polymerases.
Features of AccuPrime Taq DNA Polymerase, High Fidelity
- High specificity and yield for robust PCR amplification
- 9-fold higher fidelity than Taq DNA polymerase alone
- Mimimal optimization steps, even with non-optimized primer sets
- Efficient amplification of targets over a broad size range up to 20 kb
How it works
Pyrococcus species GB-D polymerase is a proofreading enzyme that possesses a 3' to 5' exonuclease activity. Mixing this enzyme with Taq DNA Polymerase increases fidelity and allows amplification of simple and complex DNA templates over a large range of target sizes.
The Platinum antibody complexes with Taq DNA polymerase and inhibits activity at room temperature. Activity is restored after the initial denaturation step, providing an automatic hot start PCR.
The thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, preventing mispriming and enhancing PCR specificity and yield.
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Content And Storage | • AccuPrime Taq DNA Polymerase, High Fidelity (1 x 200 μL at 5 U/μL) • 10X AccuPrime PCR Buffer I (4 x 1.25 mL) • 10X AccuPrime PCR Buffer II (4 x 1.25 mL) • 50 mM MgSO4 (4 x 1.25 mL) Store at -10°C to -30°C. |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | Low |
| Polymerase | AccuPrime Taq DNA Polymerase High Fidelity |
| Reaction Speed | Standard |
| Product Type | DNA Polymerase |
| Quantity | 1,000 reactions |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Hot-start PCR, High-fidelity PCR |
| Fidelity (vs. Taq) | 9X |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.