Learn More
Invitrogen™ 96 Deep Well Block
Description
Includes
Fifty 96 Deep Well Blocks
With 2.2mL capacity per well, 96 Deep Well Block can be used for manual sample processing or for high-throughput, automated processing using liquid handling robot.
- 96 square-diameter wells, conforming to proposed SBS microplate standards
- Conical-shaped well bottom to facilitate sample retrieval
- 2.2mL working volume per well when left unsealed or when sealed with adhesive or heat seal
- Support and stacking ribs
DNA and RNA Purification and Analysis, DNA Extraction, Genomic DNA Purification, Miniprep, PCR Product Clean-Up, Plasmid DNA Purification, gDNA from Blood (Small-Scale, < 100 μl), gDNA from Buccal Cells, gDNA from Forensic Samples, gDNA from Plant and Fungi
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Kit Contents | Fifty 96 Deep Well Blocks |
| For Use With (Application) | DNA and RNA Purification and Analysis, DNA Extraction, Genomic DNA Purification, Miniprep, PCR Product Clean-Up, Plasmid DNA Purification, gDNA from Blood (Small-Scale, < 100μL), gDNA from Buccal Cells, gDNA from Forensic Samples, gDNA from Plant and Fungi |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.