missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 25 bp DNA Ladder

Product Code. 11598626
Click to view available options
Quantity:
50 μL
Unit Size:
50µg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11598626

Brand: Invitrogen™ 10597011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Includes

25bp DNA Ladder (1μg/μL) in 10mM Tris-HCl (pH 7.5), 1mM EDTA

Suitable for sizing double-stranded DNA from 25 bp to 500 bp

Prepared from a plasmid containing repeats of a 25bp DNA fragment, the ladder consists of 19 blunt fragments ranging in length from 25 to 450bp, at 25bp increments, plus a 500bp band.

Product use

  • This ladder can be stained with ethidium bromide or radiolabeled using T4 DNA polymerase or T4 polynucleotide kinase
  • The 125bp band is approximately 2 to 3 times brighter than the other bands, except for the 500bp and 2652bp bands, to provide internal orientation
  • This ladder is not designed for quantitation. Compatible with agarose gels

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/μL
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Kit Contents 25bp DNA Ladder (1μg/μL) in 10mM Tris-HCl (pH 7.5), 1mM EDTA
Quantity 50 μL
Range 25 to 500 bp

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.