missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 123 bp DNA Ladder

Product Code. 11588756
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588756 250 μL 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11588756 Supplier Invitrogen™ Supplier No. 15613029

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

Concentration 1 μg/μL
Form Liquid
Kit Contents Supplied at 1μg/μL in 10mM Tris-HCl (pH 7.5), 50mM NaCl, 0.1mM EDTA
Quantity 250 μL
Range 123 to 3075 bp
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.