Learn More
Invitrogen™ 1kb DNA Extension Ladder
Description
Includes
1 Kb DNA Extension Ladder is supplied at 1μg/μL in 10mM Tris-HCl (pH 7.5), 50mM NaCl, 0.1mM EDTA Volume: 100μL
The 1Kb DNA Extension Ladder consists of 8 fragments ranging in length from 1,018 to 8,144bp in 1,018-bp increments, as well as bands of 506bp, 517bp, 1,636bp, 10Kb, 15Kb, 20Kb, and 40Kb. For easy reference on agarose gels, the 5Kb band is two to three-times brighter than the other bands in the ladder. This ladder can be stained with ethidium bromide or radiolabeled using T4 polynucleotide kinase, T4 DNA polymerase, DNA polymerase I, or the large fragment DNA polymerase I (Klenow fragment). Compatible with agarose gels.
- Less waste, sustainable packaging
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Concentration | 1 μg/μL |
| Form | Liquid |
| Kit Contents | 1Kb DNA Extension Ladder is supplied at 1μg/μL in 10mM Tris-HCl (pH 7.5), 50mM NaCl, 0.1mM EDTA |
| Quantity | 100 μL |
| Range | 0.5 to 40kb |
| Product Type | DNA Extension Ladder |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.