Learn More
Inventory Storage Racks
Product Code. 15377243
Change view
Click to view available options
Holds:
A-1 Tubes
Product Code. Holds
15377243 A-1 Tubes
1 options
Product Code. 15377243 Supplier PHCbi Supplier No. NUNIR209C
Only null left
Add to basket
Add to basket
These Inventory Storage Racks provide cold storage solutions in ultra-low temperatures within cryogenic freezers.
  • For use in cryogenic chest freezers MDF-1156
Specifications
Holds A-1 Tubes
Material Aluminum
Positions 9
For Use With (Equipment) Panasonic MDF-1156/ATN
Dimensions (L x W x H) 144 x 142 x 518 mm
Product Type Freezer Rack
Height (Metric) 52 mm