Learn More
Invar|r, Iron Nickel powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 044600.36
Code : NEW
Additional Details : Weight : 0.50000kg Transport : UN number : 3089 Chem class : 4.1 Pack group : III
Description
Invar is used where high dimensional stability is required, such as precision instruments, clocks, seismic creep gauges, television shadow-mask frames, valves in motors, and antimagnetic watches.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
Fe:Ni 65:35 wt% | |
MFCD01741928 | |
500 g | |
99.9% | |
Odorless |
Invar™ Iron Nickel powder | |
(metals basis) | |
UN3089 | |
Insoluble ion water. | |
−325 Mesh | |
Powder |
RUO – Research Use Only