missing translation for 'onlineSavingsMsg'
Learn More

Inulin, MP Biomedicals™

Product Code. 11446921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446921 25 g 25g
11456921 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446921 Supplier MP Biomedicals Supplier No. 0219897125
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-80-5
Molecular Formula (C6H10O5)n
Synonym Dahlin

Specifications

Color White
Quantity 25 g
Formula Weight ≈5,000
Physical Form Free-flowing Powder
Chemical Name or Material Inulin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.