missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11657-1-AP-150UL
This item is not returnable.
View return policy
Description
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Specifications
| INTS9 | |
| Polyclonal | |
| Unconjugated | |
| INTS9 | |
| CPSF2L, INT9, integrator complex subunit 9, INTS9, RC 74, RC74 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55756 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NV88 | |
| INTS9 | |
| INTS9 Fusion Protein Ag2226 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction