missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
INTS6 Polyclonal specifically detects INTS6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | INTS6 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | DBI1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26, DICE1DBI-1DDX26ADDX26DEAD box protein, DKFZp434B105, HDB, INT6, integrator complex subunit 6, Notchl2, Protein DDX26, Protein deleted in cancer 1, RNA helicase HDB |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human INTS6 (NP_036273.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?