missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INTS6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551643
This item is not returnable.
View return policy
Description
Immunogen sequence: VVNNHIGGKG PPAPTTQAQP DLIKPLPLHK ISETTNDSII HDVVENHVAD QLSSDITPNA MDTEFSASSP ASLLERPTNH MEALGHDHLG TNDLTVGGFL ENHEEPRDKE QCAEENIPAS SLNKGKKLMH CRSHEEVNTE LKAQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. INTS6 is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. The protein encoded by this gene is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| INTS6 | |
| Polyclonal | |
| Unconjugated | |
| INTS6 | |
| 2900075H24Rik; AI480962; Dbi1; DBI-1; Ddx26; DDX26A; DEAD box protein; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26; DICE1; DKFZP434B105; HDB; INT6; integrator complex subunit 6; INTS6; LRRGT00024; Notch2l; Notchl2; Protein DDX26; Protein deleted in cancer 1; RNA helicase HDB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UL03 | |
| INTS6 | |
| Recombinant protein corresponding to Human INTS6. Recombinant protein control fragment (Product #RP-95936). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction