missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS5 Polyclonal Antibody

INTS5 Polyclonal Antibody
Brand: Invitrogen PA5121017
This item is not returnable.
View return policy
Description
INTS5 Polyclonal Antibody for Western Blot
Specifications
| INTS5 | |
| Polyclonal | |
| Unconjugated | |
| INTS5 | |
| 1110055N21Rik; INT5; integrator complex subunit 5; Ints5; KIAA1698; mKIAA1698; RGD1309370 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109077, 80789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P9B9, Q8CHT3 | |
| INTS5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human INTS5 (NP_085131.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction