Learn More
Invitrogen™ INTS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109865
Description
Immunogen sequence: DVVQEVQQVL SEFIRANPKA WAPVISAWSI DLMGQLSSTY SGQHQRVPHA TGALNELLQL WMGCRATRTL MDIYVQCLSA LIGSCPDACV DALLDTSVQH SPHFDWVVAH IGSSFPGTII SRVLSCGLKD FCVHGGAGGG AGSSGGSSSQ TPSTDPFPGS PAIPAEKRVP KIASVVGILG HLASRHGDSI RRELLRMFHD SLAGGSGGRS GDPSLQATVP FLLQLAVMSP ALLGTVSGEL VDCLKPPAVL SQLQQHLQGF PREELDNMLN LAVHLVSQAS G.
INTS5 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
Specifications
| INTS5 | |
| Polyclonal | |
| Unconjugated | |
| INTS5 | |
| 1110055N21Rik; INT5; integrator complex subunit 5; Ints5; KIAA1698; mKIAA1698; RGD1309370 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109077, 80789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P9B9, Q8CHT3 | |
| INTS5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human INTS5 (NP_085131.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.