missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTS3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567343

Product Code. 15960005

  • £471.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LEHLDNLRLNL TNTKQNFFSQ TPILQALQHV QASCDEAHKM KFSDLFSLAE EYEDSSTKPP KSRRKAALSS PRSRKNATQP Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.

Component of the Integrator complex. The Integrator complex is involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing. The Integrator complex is associated with the C-terminal domain (CTD) of RNA polymerase II largest subunit (POLR2A) and is recruited to the U1 and U2 snRNAs genes.; FUNCTION: Component of the SOSS complex, a multiprotein complex that functions downstream of the MRN complex to promote DNA repair and G2/M checkpoint. The SOSS complex associates with single-stranded DNA at DNA lesions and influences diverse endpoints in the cellular DNA damage response including cell-cycle checkpoint activation, recombinational repair and maintenance of genomic stability. The SOSS complex is required for efficient homologous recombination-dependent repair of double-strand breaks (DSBs) and ATM-dependent signaling pathways. In the SOSS complex, it is required for the assembly of the complex and for stabilization of the complex at DNA damage sites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INTS3
Polyclonal
Unconjugated
INTS3
C1orf193; C1orf60; C77668; INT3; integrator complex subunit 3; Ints3; RGD1304593; Sensor of single-strand DNA complex subunit A; sensor of ssDNA subunit A; SOSS complex subunit A; SOSSA; SOSS-A
Rabbit
Antigen affinity chromatography
RUO
65123
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q68E01
INTS3
Recombinant Human INTS3. Recombinant protein control fragment (Product #RP-107993).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato