missing translation for 'onlineSavingsMsg'
Learn More

INTS11 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£161.00 - £383.00

Specifications

Antigen INTS11
Dilution Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30226888
View Documents
Novus Biologicals
NBP3-33376-100ul
100 μL
£383.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227814
View Documents
Novus Biologicals
NBP3-33376-20ul
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INTS11 Monoclonal antibody specifically detects INTS11 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

INTS11
ELISA, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
cleavage and polyadenylation specific factor 3-like, Cleavage and polyadenylation-specific factor 3-like protein, CPSF3-like protein, EC 3.1.27, EC 3.1.27.-, FLJ20542, INT11, integrator complex subunit 11, INTS11FLJ13294, Protein related to CPSF subunits of 68 kDa, RC68, RC-68CPSF73L, related to CPSF subunits 68 kDa
Recombinant fusion protein containing a sequence corresponding to amino acids 331-600 of human INTS11 (NP_060341.2).,, Sequence:, GQSLQIFRKWAGNEKNMVIMPGYCVQGTVGHKILSGQRKLEMEGRQVLEVKMQVEYMSFSAHADAKGIMQLVGQAEPESVLLVHGEAKKMEFLKQKIEQELRVNCYMPANGETVTLPTSPSIPVGISLGLLKREMAQGLLPEAKKPRLLHGTLIMKDSNFRLVSSEQALKELGLAEHQLRFTCRVHLHDTRKEQETALRVYSHLKSVLKDHCVQHLPDGSVTVESVLLQAAAPSEDPGTKVLLVSWTYQDEELGSFLTSLLKKGLPQAPS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol, 0.05% BSA
54973
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.