missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INTS10 Recombinant Rabbit Monoclonal Antibody (K01_3K22)
Description
INTS10 Recombinant Monoclonal Antibody for Western Blot
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1.
Specifications
Specifications
| Antigen | INTS10 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_3K22 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | Ints10 |
| Gene Accession No. | Q8K2A7, Q9NVR2 |
| Gene Alias | 4921521J11Rik; AI462004; C8orf35; int10; Integrator complex subunit 10; INTS10; RGD1304711 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?