missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15271-1-AP-150UL
This item is not returnable.
View return policy
Description
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1.Specifications
| INTS10 | |
| Polyclonal | |
| Unconjugated | |
| INTS10 | |
| C8orf35, INT10, integrator complex subunit 10, INTS10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290679, 55174, 70885 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K2A7, Q9NVR2 | |
| Ints10 | |
| INTS10 Fusion Protein Ag7383 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction