Learn More
INTS10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15271-1-AP-20UL
 View more versions of this product
Product Code. 16834374
  • £135.00 / 20µL
Description
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1.
Specifications
Specifications
INTS10
Polyclonal
Unconjugated
INTS10
C8orf35, INT10, integrator complex subunit 10, INTS10
Rabbit
Antigen Affinity Chromatography
RUO
290679, 55174, 70885
-20°C
Liquid
Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K2A7, Q9NVR2
Ints10
INTS10 Fusion Protein Ag7383
20 μL
Primary
Rat, Human, Mouse