missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTS10 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589225

Product Code. 16587191

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FLTDMIIYQG QYKKAIASLH HLAALQGSIS QPQITGQGTL EHQRALIQLA TCHFALGEYR MTCEKVLDLM CYMVLPIQDG GKSQEEPSKV KPKFRKGSDL KLLPCTSKAI MPYCLHLMLA CFKLRAFTDN RDDMALGHVI VLLQQEWPRG ENLFLKAVNK ICQQGNFQYE NFFNYVTNID MLEEFAYLRT QEGGKIHLEL LPNQGMLIKH HTVTRGITKG VKEDFRLAME RQVSRCGENL MVVLHRFCIN EKILLLQTLT; Positive Samples: A-549, HL-60, HeLa, HepG2, Mouse brain; Cellular Location: Nucleus.

INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INTS10
Polyclonal
Unconjugated
Ints10
4921521J11Rik; AI462004; C8orf35; int10; Integrator complex subunit 10; INTS10; RGD1304711
Rabbit
Affinity Chromatography
RUO
55174, 70885
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K2A7, Q9NVR2
Ints10
Recombinant fusion protein containing a sequence corresponding to amino acids 451-710 of human INTS10 (NP_0606122).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.