missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intra Acrosomal Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34054-25ul
This item is not returnable.
View return policy
Description
Intra Acrosomal Protein Polyclonal specifically detects Intra Acrosomal Protein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Intra Acrosomal Protein | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P26436 | |
| ACRV1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TSSQPNELSGSIDHQTSVQQLPGEFFSLENPSDAEALYETSSGLNTLSEHGSSEHGSSKHTVAEHTSGEHAE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction