missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Intra Acrosomal Protein Polyclonal specifically detects Intra Acrosomal Protein in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Intra Acrosomal Protein |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Intra Acrosomal Protein (P26436-1; Gln22-Ile265). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?