missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intra Acrosomal Protein Antibody (Ds-2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-436AF750
This item is not returnable.
View return policy
Description
Intra Acrosomal Protein Monoclonal specifically detects Intra Acrosomal Protein in Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Intra Acrosomal Protein | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ACRV1 | |
| Freshly ejaculated dog sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid, lyophylized and subsequently used for immunization. | |
| 0.1 mL | |
| 56.0 | |
| Canine | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Ds-2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The antibody Ds-2 reacts with canine intra-acrosomal sperm protein (92 and 42 kDa), a testis-specific acrosomal vesicle protein 1 associated with the acrosomal membranes and matrix of the mature sperm. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction