missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intestinal Alkaline Phosphatase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA580334
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ALPI/IAP.
AP is an enzyme primarially localized in plasma membranes. It can remove terminal phosphates on several types of molecules and thus it is an important tool for biological research. When alkaline phosphatase is coupled to streptavidin and immobilized to the solid phase in ELISA, HYB 252-08-02 binds strongly to the presented alkaline phosphatase.
Specifications
| Intestinal Alkaline Phosphatase | |
| Polyclonal | |
| PBS with no preservative | |
| P09923 | |
| ALPI | |
| Recombit Human ALPI/IAP protein | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| ALPI | |
| AP, Akp2, Alkaline phosphatase intestinal, Alkaline phosphomonoesterase, ALPI, Glycerophosphatase, IAP, Kasahara isozyme | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 248 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction