missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intersectin 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71832AF488
This item is not returnable.
View return policy
Description
Intersectin 1 Polyclonal antibody specifically detects Intersectin 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Intersectin 1 | |
| Polyclonal | |
| Western Blot | |
| 11, intersectin 1 (SH3 domain protein), intersectin short variant 12, intersectin-1, ITSN, SH3 domain protein-1A, SH3 domain-containing protein 1A, Src homology 3 domain-containing protein | |
| The immunogen for this product maps to a region between residue 850 and 900 of human Intersectin 1 using the numbering given in entry NP_003015.2 (GeneID 6453). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6453 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction