missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Internal Standards Mixture 508 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobenzene-D4∣Chrysene D12∣Phenanthrene D10 |
| CAS | 3855-82-1, 1719-03-5, 1517-22-2 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Dichlorobenzene-D4 100 μg/mL∣Chrysene D12 100 μg/mL∣Phenanthrene D10 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction