missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Internal Standard Multi-Element Mix 3: 6Li, Sc, Ge, In, Tb, Lu, Bi @ 100 μg/mL in 5% HNO3, tr. F-, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Internal Standard Multi-Element Mix 3: 6Li, Sc, Ge, In, Tb, Lu, Bi @ 100 μg/mL in 5% HNO3, tr. F-
Specifications
Specifications
| Chemical Name or Material | Bismuth/Germanium/Indium/Lithium 6/Lutetium/Scandium/Terbium |
| CAS | 7440-69-9, 7440-56-4, 7440-74-6, 14258-72-1, 7439-94-3, 7440-20-2 |
| Methods | ICP-MS |
| Analyte or Component Names | Bismuth/Germanium/Indium/Lithium 6/Lutetium/Scandium/Terbium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Bismuth 100 μg/mL/Germanium 100 μg/mL/Indium 100 μg/mL/Lithium 6 100 μg/mL/Lutetium 100 μg/mL/Scandium 100 μg/mL/Terbium 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction