missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard: Ge @ 10 μg/mL in 2% HNO3, tr. F-, VHG™

Product Code. 17951496
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
17951496 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 17951496

Brand: VHG Labs VHGLISAGE100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Internal Standard: Ge @ 10 μg/mL in 2% HNO3, tr. F-
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Germanium
CAS 7440-56-4
Methods ICP-MS
Analyte or Component Names Germanium
Shipping Condition Room Temperature
Molecular Formula Ge
Certifications/Compliance ISO 17034
Synonym Ge
InChI Formula InChI=1S/Ge
SMILES [Ge]
IUPAC Name germanium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 72.64
Quantity 100 mL
Formula Weight 73.9212 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.