Learn More
Manutan Internal doors, quick hang PVC curtains, 3200mm x 2000mm
Product Code. 12874529
Encompass_Preferred
Product Code. 12874529 Supplier Manutan Supplier No. A352121
This item requires a price update prior to being ordered.
Request a Price for this item here.
  • Drive through flexible curtains for internal doors. PVC strips create a see through flexible curtain. Keeps heat in and reduces cold air loss in chill/freezer rooms for greater temperature control. Provides protection against rain, wind and restricts insect and bird access. Forms a good noise barrier with partial (50%) overlap.
  • Dimensions (H x W): 3200mm x 2000mm