missing translation for 'onlineSavingsMsg'
Learn More

Interferon receptor inducer-1, MedChemExpress

Product Code. 16460897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460897

Brand: Medchem Express HY112189

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Interferon receptor inducer-1 (compound 6) is an interferon (IFN) receptor inducer. Used accordingly in the treatment of a disorder in which the induction of interferon is involved.

Interferon receptor inducer-1 (compound 6) is an interferon (IFN) receptor inducer. Used accordingly in the treatment of a disorder in which the induction of interferon is involved.

Specifications

Chemical Name or Material Interferon receptor inducer-1
Physical Form Solid
CAS 2215120-36-6
Color Pink
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H22N4O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 55 mg/mL (216.25 mM)
SMILES CCCC[C@H](CO)NC1=C(OC)C=NC(NC)=N1
Molecular Weight (g/mol) 254.33
Formula Weight 254.33
Percent Purity 99.15%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.