missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon alpha-4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535276
This item is not returnable.
View return policy
Description
Interferon alpha-4 Polyclonal Antibody for Western Blot, IHC (P)
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Interferon alpha-4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P05014 | |
| IFNA4 | |
| KLH conjugated synthetic peptide between 162-189 amino acids from the C-terminal region of human IFNA4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| IFNA4 | |
| Ifa4; IFNA4; IFN-alpha-4; IFN-alpha4a; INFA4; Interferon; interferon alpha 4; interferon alpha family, gene 4; interferon alpha-4; Interferon alpha-4B; interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction