missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon alpha-21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595848
This item is not returnable.
View return policy
Description
Immunogen sequence: WEQSLLEKFS TELNQQLNDL EACVIQEVGV EETPLMNVDS ILAVKKYFQR ITLYLTEKKY SPCAWEVVRA EIMRSFSLSK IFQERLRRKE.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Interferon alpha-21 | |
| Polyclonal | |
| Unconjugated | |
| IFNA21; IFNA5; IFNA6; IFN-alpha-21; IFN-alpha-5; IFN-alpha-6; IFN-alphaG; IFN-alphaI; IFN-alphaK; INA5; INFA5; Interferon; interferon alpha 21; interferon alpha 5; interferon alpha 6; interferon alpha-21; Interferon alpha-5; interferon alpha-54; Interferon alpha-6; interferon alpha-61; Interferon alpha-F; interferon alpha-G; Interferon alpha-K; interferon, alpha 21; interferon, alpha 5; interferon, alpha 6; LeIF F; LeIF G; LeIF K; leIF-F; leukocyte interferon protein | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human IFNA21 (NP_0021662). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.16 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction