missing translation for 'onlineSavingsMsg'
Learn More

Interference Check Standard AB(ICS-AB) Solution, Specpure™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 041730.AE

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11328888

  • £196.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Interference Check Standard AB(ICS-AB) Solution
100 mL
UN3264
Liquid
Matrix: 5% HNO3/tr. tartaric acid
MFCD00147272
Specpure™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only