missing translation for 'onlineSavingsMsg'
Learn More

Intepirdine, MedChemExpress

Product Code. 16446038
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446038

Brand: Medchem Express HY14339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Intepirdine (SB742457) is a highly selective 5-HT6 receptor antagonist with pKi of 9.63; exhibits >100-fold selectivity over other receptors.

Intepirdine (SB742457) is a highly selective 5-HT6 receptor antagonist with pKi of 9.63

Specifications

Chemical Name or Material Intepirdine
CAS 607742-69-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H19N3O2S
Quantity 10 mg
Synonym SB-742457 GSK-742457 RVT-101
Solubility Information DMSO : 50 mg/mL (141.47 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(C2=CC(C=CC=C3N4CCNCC4)=C3N=C2)=O
Molecular Weight (g/mol) 353.44
Formula Weight 353.44
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.