missing translation for 'onlineSavingsMsg'
Learn More

Intepirdine, MedChemExpress

Product Code. 16456446
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456446 100 mg 100mg
16452177 50 mg 50mg
16446038 10 mg 10mg
16408647 5 mg 5mg
16403426 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16456446 Supplier Medchem Express Supplier No. HY14339
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Intepirdine (SB742457) is a highly selective 5-HT6 receptor antagonist with pKi of 9.63; exhibits >100-fold selectivity over other receptors.

Intepirdine (SB742457) is a highly selective 5-HT6 receptor antagonist with pKi of 9.63

Specifications

Chemical Name or Material Intepirdine
CAS 607742-69-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H19N3O2S
Quantity 100 mg
Synonym SB-742457 GSK-742457 RVT-101
Solubility Information DMSO : 50 mg/mL (141.47 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(C2=CC(C=CC=C3N4CCNCC4)=C3N=C2)=O
Molecular Weight (g/mol) 353.44
Formula Weight 353.44
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.