missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTELLIFLEX Calibration Kit

Product Code. 18088051
Change view
Click to view available options
Quantity:
20 reactions kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18088051 20 reactions kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18088051 Supplier Invitrogen™ Supplier No. IFXCALK20
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems.

The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems. The calibration kit contains all reagents needed for calibration of these systems.

Calibration is important to ensure that the optical system functions effectively and that different INTELLIFLEX systems report similar results. Calibrating the INTELLIFLEX System normalizes the settings for classification channels (CL1, CL2, and CL3), the doublet discriminator channel (DD), and the reporter channels (RP1 and RP2).

Please refer to the INTELLIFLEX Calibration Kit package insert for proper use of the INTELLIFLEX Calibration Kit.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Luminex™ xMAP™ INTELLIFLEX, Luminex™ xMAP™ INTELLIFLEX DR-SE
No. of Tests 20 Calibrations
Quantity 20 reactions kit
Content And Storage • xMAP INTELLIFLEX Calibrator, 10 mL
• 25 Strips wells

Each kit will provide 20 calibrations.
Store in the dark at 2°C to 8°C.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.