missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intelectin-1/Omentin Mouse anti-Human, Clone: ITLN1/4066, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Intelectin-1/Omentin |
|---|---|
| Clone | ITLN1/4066 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Intelectin-1/Omentin | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin | |
| Recombinant fragment (around aa16-141) of human Intelectin-1/Omentin protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| ITLN1/4066 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 55600 | |
| IgG2c κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title