missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 8 Antibody (CL7290) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76480B
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Integrin beta 8 | |
| Monoclonal | |
| Biotin | |
| integrin beta-8, integrin, beta 8 | |
| This antibody was developed using a recombinant protein derived from P26012, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 3696 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL7290 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction