missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Integrin beta 8 Antibody (2B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00003696-M01
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay
Specifications
| Integrin beta 8 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| integrin beta-8, integrin, beta 8 | |
| ITGB8 (NP_002205, 392 a.a. ~ 503 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VENQVQGIYFNITAICPDGSRKPGMEGCRNVTSNDEVLFNVTVTMKKCDVTGGKNYAIIKPIGFNETAKIHIHRNCSCQCEDNRGPKGKCVDETFLDSKCFQCDENKCHFDE | |
| 0.1 mg | |
| Signal Transduction | |
| 3696 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay | |
| 2B4 | |
| Western Blot 1:500, Flow Cytometry, ELISA, Immunoprecipitation, Functional | |
| NP_002205 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion