missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 7 Antibody, R&D Systems™
Rat Monoclonal Antibody
£333.68
Specifications
Antigen | Integrin beta 7 |
---|---|
Clone | 332428 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Western Blot.Specifications
Integrin beta 7 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P26011 | |
3695 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 7 Glu20-His722 Accession # P26011 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Integrin beta 7 extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Integrin beta 1, beta 2, beta 3, beta 4, beta 5, or beta 6 is observed. |
332428 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7, ITGB7 | |
ITGB7 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only