missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (Genentech patent anti-Integrin beta7), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28000MFV500
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Integrin b7/ ITGB7 | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-Integrin beta7 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction