missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (FIB504), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81086PE
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| PE | |
| PBS | |
| ITGB7 | |
| FIB504 was prepared by immunizing rats with TK1 cells. | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| FIB504 | |
| Flow Cytometry | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| FIB504 recognizes an epitope on Integrin beta 7 subunit that overlaps completely with the epitope defined by Fib27 and Fib30, and also partially with the epitope defined by FIB21 and FIB22. In the mouse beta-7 is selectively found on the majority of mature lymphocytes, whereas a small subpopulation of thymocytes and bone marrow cells express beta-7. Beta-7 is also expressed in variable amounts in CD4 T memory cells. Integrin molecules mediate cell-cell and cell-extracellular matrix adhesion and are involved in lymphocyte homing, leukocyte recruitment to inflammatory sites, myogenesis, hemopoiesis, and melanoma metastasis. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction