missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7 Antibody (FIB504), DyLight 594, Novus Biologicals™

Product Code. 18332111 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332111 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332111 Supplier Novus Biologicals Supplier No. NBP281086DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 7
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FIB504
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Gene Symbols ITGB7
Host Species Rabbit
Immunogen FIB504 was prepared by immunizing rats with TK1 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3695
Test Specificity FIB504 recognizes an epitope on Integrin beta 7 subunit that overlaps completely with the epitope defined by Fib27 and Fib30, and also partially with the epitope defined by FIB21 and FIB22. In the mouse beta-7 is selectively found on the majority of mature lymphocytes, whereas a small subpopulation of thymocytes and bone marrow cells express beta-7. Beta-7 is also expressed in variable amounts in CD4 T memory cells. Integrin molecules mediate cell-cell and cell-extracellular matrix adhesion and are involved in lymphocyte homing, leukocyte recruitment to inflammatory sites, myogenesis, hemopoiesis, and melanoma metastasis.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.