missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7 Antibody (332418), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals FAB3060J

405.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18553768

  • £462.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Integrin beta 7
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
332418
Flow Cytometry, CyTOF-ready
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 7, Glu20-His722, Accession # P26011
0.1 mL
Signal Transduction
3695
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.