missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 6 Antibody (CSbeta6), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18427336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427336 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427336 Supplier Novus Biologicals Supplier No. NBP229800AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Human, Rat, Primate, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 6
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CSbeta6
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias integrin beta-6, integrin, beta 6
Host Species Mouse
Immunogen beta 6 knockout mice have been immunized with recombinant secreted human alpaVbeta6.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694
Target Species Human, Rat, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.