missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Integrin beta 6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 437216 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | integrin beta-6, integrin, beta 6 |
| Host Species | Mouse |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Integrin β6, Gly22-Asn707, Accession # P18564 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?