missing translation for 'onlineSavingsMsg'
Learn More

integrin beta 4 binding protein Antibody, DyLight 488, Novus Biologicals™

Product Code. 18401933 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401933 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401933 Supplier Novus Biologicals Supplier No. NBP152641G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

integrin beta 4 binding protein Polyclonal antibody specifically detects integrin beta 4 binding protein in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen integrin beta 4 binding protein
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias b(2)gcn, B(2)GCN homolog, B4 integrin interactor, CAB, EIF3Ap27(BBP), eIF-6, eukaryotic translation initiation factor 3A, eukaryotic translation initiation factor 6, ITGB4BPintegrin beta 4 binding protein, p27 beta-4 integrin-binding protein, p27BBP
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Eukaryotic Translation Initiation Factor 6 using the numbering given in entry NP_852133.1 (GeneID 3692).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3692
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.