missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Rabbit anti-Human, Clone: 60.3, Novus Biologicals™
Rabbit Monoclonal Antibody
£210.00 - £463.00
Specifications
| Antigen | Integrin beta 2/CD18 |
|---|---|
| Clone | 60.3 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Functional (Inhibition), Block/Neutralize |
| Applications | Western Blot, ELISA, Immunofluorescence, Inhibition Assays, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335382
|
Novus Biologicals
NBP3-12031-0.025MG |
0.025 mg |
£210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375702
|
Novus Biologicals
NBP3-12031-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Inhibition Assays, NeutralizationSpecifications
| Integrin beta 2/CD18 | |
| 1 mg/ml | |
| Western Blot, ELISA, Immunofluorescence, Inhibition Assays, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
| PBS | |
| 3689 | |
| IgG κ | |
| Protein A purified |
| 60.3 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Functional (Inhibition), Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| The original antibody was generated by immunizing BALB/c mice with alloantigen activated T cells. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title