missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 2/CD18 Antibody (60.3), DyLight 594, Novus Biologicals™

Product Code. 18372113 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372113 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372113 Supplier Novus Biologicals Supplier No. NBP312031DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 2/CD18
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 60.3
Conjugate DyLight 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Host Species Rabbit
Immunogen The original antibody was generated by immunizing BALB/c mice with alloantigen activated T cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.