Learn More
Integrin beta 2/CD18 Antibody (1B4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09040-0.025mg
 View more versions of this product
Product Code. 18632389
  • £206.00 / 25µg
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human, Macaque samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin beta 2/CD18
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Macaque
Purified
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA)
1B4
Unconjugated
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Blood was obtained from 4 helathy donors and the mononuclear cells harvested and grown. A BALB/c mouse was injected with 10^6 monocytes three times at monthly intervals, and four times with 5x10^6 monocytes oon each of the four days before fusion. The spleen cells were then fused to P3U1 myeloma cells to generate hybridomas and RIA was used to identify lines of interest.
0.025 mg
Apoptosis, Cytokine Research, Immunology
3689
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG κ